missing translation for 'onlineSavingsMsg'
Learn More

Linear Eptifibatide TFA Salt(Reduced), TRC

Product Code. 30478100
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478100 1 mg 1mg
30477081 5 mg 5mg
30472908 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478100 Supplier Toronto Research Chemicals Supplier No. TRCL4664801MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Linear Eptifibatide TFA Salt(Reduced)
Analyte or Component Names Linear Eptifibatide TFA Salt(Reduced)
Shipping Condition Room Temperature
Molecular Formula C35H51N11O9S2. x C2F3OOH
InChI Formula InChI=1S/C35H51N11O9S2.C2HF3O2/c36-30(51)25(18-57)45-33(54)26-9-5-12-46(26)34(55)24(14-19-16-40-21-7-2-1-6-20(19)21)44-32(53)23(15-29(49)50)43-28(48)17-41-31(52)22(42-27(47)10-13-56)8-3-4-11-39-35(37)38;3-2(4,5)1(6)7/h1-2,6-7,16,22-26,40,56-57H,3-5,8-15,17-18H2,(H2,36,51)(H,41,52)(H,42,47)(H,43,48)(H,44,53)(H,45,54)(H,49,50)(H4,37,38,39);(H,6,7)/t22-,23-,24+,25+,26-;/m0./s1
Solution Type Neat
SMILES SCCC(N[C@@H](CCCCNC(N)=N)C(NCC(N[C@@H](CC(O)=O)C(N[C@H](CC1=CNC2=C1C=CC=C2)C(N(CCC3)[C@@H]3C(N[C@@H](C(N)=O)CS)=O)=O)=O)=O)=O)=O.FC(F)(C(O)=O)F
Industry Type Pharmaceutical
Molecular Weight (g/mol) 948
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.