missing translation for 'onlineSavingsMsg'
Learn More

Linomide

Product Code. 15436909
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436909 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436909 Supplier Thermo Scientific Alfa Aesar Supplier No. J65453.MA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84088-42-6 | C18H16N2O3 | 308.337 g/mol

An immunomodulator with antitumour properties. Inhibits T-cell apoptosis; also inhibits angiogenesis in rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunomodulator with antitumour properties
  • Inhibits T-cell apoptosis; also inhibits angiogenesis in rats
TRUSTED_SUSTAINABILITY

Specifications

CAS 84088-42-6
Color White
Molecular Formula C18H16N2O3
MDL Number MFCD00866331
Quantity 10 mg
Synonym linomide, roquinimex, roquinimexum latin, roquinimex usan:inn, ccris 7673, fcf 89, fcf-89, n-phenyl-n-methyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-quinoline-3-carboxamide, 1,2-dihydro-4-hydroxy-n,1-dimethyl-2-oxo-3-quinolinecarboxanilide, 4-hydroxy-n,1-dimethyl-2-oxo-n-phenyl-1,2-dihydroquinoline-3-carboxamide
InChI Key SGOOQMRIPALTEL-UHFFFAOYSA-N
SMILES CN1C2=CC=CC=C2C(=C(C1=O)C(=O)N(C)C3=CC=CC=C3)O
IUPAC Name 4-hydroxy-N,1-dimethyl-2-oxo-N-phenylquinoline-3-carboxamide
Molecular Weight (g/mol) 308.337
PubChem CID 54676478
Formula Weight 308.33
Physical Form Solid
Chemical Name or Material Linomide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.