missing translation for 'onlineSavingsMsg'
Learn More

Lintopride, MedChemExpress

Product Code. 16450696
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16450696

Brand: Medchem Express HYU00121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lintopride is a 5HT4 antagonist with moderate 5HT3 antagonist properties.

Lintopride is a 5HT4 antagonist with moderate 5HT3 antagonist properties.

Specifications

Chemical Name or Material Lintopride
CAS 107429-63-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H19ClN4O2
Quantity 20 mg
SMILES O=C(NCC1=NCCN1CC)C2=CC(Cl)=C(N)C=C2OC
Molecular Weight (g/mol) 310.78
Formula Weight 310.78
Percent Purity 96.38%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.