missing translation for 'onlineSavingsMsg'
Learn More

Liothyronine, MedChemExpress

Product Code. 16478016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478016 100 mg 100mg
16481976 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478016 Supplier Medchem Express Supplier No. HYA0070A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Liothyronine is an active form of thyroid hormone. Liothyronine is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Liothyronine is an active form of thyroid hormone. Liothyronine is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.

Specifications

Chemical Name or Material Liothyronine
CAS 6893-02-3
Color White
Purity Grade Notes Research
Molecular Formula C15H12I3NO4
Quantity 100 mg
Synonym Triiodothyronine 3,3',5-Triiodo-L-thyronine T3
Solubility Information DMSO : 50 mg/mL (76.81 mM; Need ultrasonic) ∣0.1 M NaOH : 16.67 mg/mL (25.61 mM; ultrasonic and warming and adjust pH to 11 with NaOH and heat to 60°C)
SMILES OC1=C(I)C=C(OC2=C(I)C=C(C[C@H](N)C(O)=O)C=C2I)C=C1
Molecular Weight (g/mol) 650.97
Formula Weight 650.97
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.