missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIPI Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149089
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Hydrolyzes specifically phosphatidic acid (PA) to produce 2-acyl lysophosphatidic acid (LPA; a potent bioactive lipid mediator) and fatty acid. Does not hydrolyze other phospholipids, like phosphatidylserine (PS), phosphatidylcholine (PC) and phosphatidylethanolamine (PE) or triacylglycerol (TG).
Specifications
| LIPI | |
| Polyclonal | |
| Unconjugated | |
| LIPI | |
| cancer/testis antigen 17; CT17; D930038D03Rik; lipase I; lipase member I; lipase, member I; LIPI; LPD lipase; lpd1; LPDL; Membrane-associated phosphatidic acid-selective phospholipase A1-beta; mPA-PLA1 beta; PLA1C; PRED5 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 149998 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6XZB0 | |
| LIPI | |
| E. coli-derived recombinant human Lipase I, Arg14-His154. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction