Learn More
Lipocalin-2/NGAL Mouse anti-Human, Clone: 14, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-41365-0.025MG
 View more versions of this product
Product Code. 18121708
  • 226.00 € / 25µg
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lipocalin-2/NGAL
Monoclonal
1 mg/ml
ELISA 1:100-1:2000
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Human recombinant NGAL coupled to carrier.
0.025 mg
Lipid and Metabolism
3934
Store at 4°C in the dark.
IgG2a κ
ELISA
14
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl
Mouse
Protein A or G purified
RUO
Primary
Human
Purified