missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 11935109
Click to view available options
Quantity:
1 kg
5 kg
546-89-4
C2H3LiO2
65.98
MFCD00013057
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
3474584
CHEBI:63045
lithium;acetate
[Li+].CC([O-])=O
546-89-4
C2H3LiO2
65.98
MFCD00013057
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
3474584
CHEBI:63045
lithium;acetate
[Li+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280.0°C to 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula CH3COOLi
Quantity 1 kg
Beilstein 02,107
Merck Index 15,5577
Solubility Information Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C),938g/L (50°C),freely soluble in alcohol
Formula Weight 65.99
Percent Purity 99%
Grade Extra Pure
Insoluble Matter 0.1% max. (in water)
Physical Form Crystals or Powder
Chemical Name or Material Lithium acetate, anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Lithium acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'