missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetylide ethylenediamine complex, tech. 85%, Thermo Scientific Chemicals

Product Code. 11456780
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11456780

Brand: Thermo Scientific Alfa Aesar L12401.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

Solubility
Soluble in ethylene diamine, n-propyl amine, n-butyl amine, pyridine, dimethyl formamaide, diglyme, dioxan and dimethylsulfoxide.

Notes
Moisture and light sensitive. Store in cool place. Incompatible with alcohols, acids, oxidizing agents, water. It reacts with water to form lithium hydroxide and acetylene, which is a flammable gas. Over time, pressure may increase causing containers to burst. Handle and open container with care.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6867-30-7
Molecular Formula C4H9LiN2
Molecular Weight (g/mol) 92.07
MDL Number MFCD00013183
InChI Key QJQWXTYPTBEPGS-UHFFFAOYSA-N
Synonym lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
PubChem CID 2724010
IUPAC Name lithium;ethane-1,2-diamine;ethyne
SMILES [Li]C#C.NCCN

Specifications

Melting Point ∽76°C (decomposition)
Density 1.04
Odor Amine-like
Linear Formula HC≡CLi·NH2CH2CH2NH2
Quantity 25 g
UN Number UN3131
Beilstein 3593824
Sensitivity Moisture sensitive
Solubility Information Soluble in ethylene diamine,n-propyl amine,n-butyl amine,pyridine,dimethyl formamaide,diglyme,dioxan and dimethylsulfoxide.
Formula Weight 92.07
Percent Purity 90%
Grade Technical
Chemical Name or Material Lithium acetylide ethylenediamine complex
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.