missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium aluminum hydride, 15% solution in toluene/THF, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Packungsgröße:
800mL
Beschreibung
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagent
Spezifikation
Spezifikation
| Color | Turbid solution |
| Linear Formula | LiAlH4 |
| Concentration | ca 15%, exact strength on the certificate of analysis |
| Density | 0.8900g/mL |
| Formula Weight | 37.95 |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 0.89 |
| Quantity | 800 mL |
| Flash Point | −20°C |
| Chemical Name or Material | Lithium Aluminum hydride, 15% solution in toluene/THF |
Sicherheit und Handhabung
Produktidentifikator
- Lithium aluminium hydride, 15% solution in toluene/THF
- Danger
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur