missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™

Product Code. 10359372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10359372 100 mL AcroSeal™ Glass bottle 100mL
10575843 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10359372 Supplier Thermo Scientific Acros Supplier No. 199511000
  • ONLINE EXCLUSIVE PRICE
    75.20 €
    73.00 € / 100mL
    Save 2.20 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Aluminum hydride
Color Colorless
Name Note 4.0M Solution in Diethyl Ether
CAS 16853-85-3, 60-29-7
Molecular Formula AlH4Li
Linear Formula LiAIH4
MDL Number MFCD00011075
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Density 0.7100g/mL
Merck Index 15, 344
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
Solubility Information Solubility in water: vigorous reaction.
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES [Li+].[AlH4-]
IUPAC Name lithium(1+) alumanuide
Molecular Weight (g/mol) 37.95
PubChem CID 21226445
Formula Weight 37.95
Percent Purity 18.0 to 22.0% (as LiAlH4)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.71
Quantity 100 mL
Flash Point −017°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium aluminium hydride, 4.0M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.