missing translation for 'onlineSavingsMsg'
Learn More

Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™

Product Code. p-153953 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10174680

Brand: Thermo Scientific Acros 268831000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 4111-54-0 | C6H14LiN | 107.125 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4111-54-0, 142-82-5, 109-99-9, 100-41-4, 108-18-9
Molecular Formula C6H14LiN
Molecular Weight (g/mol) 107.125
MDL Number MFCD00064449
InChI Key ZCSHNCUQKCANBX-UHFFFAOYSA-N
Synonym lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli
PubChem CID 2724682
IUPAC Name lithium;di(propan-2-yl)azanide
SMILES [Li+].CC(C)[N-]C(C)C

Specifications

Color Brown-Red or Orange to Yellow
Boiling Point 60.0°C to 136.0°C
Composition Solvent: THF 59%, n-Heptane: 28%, Ethylbenzene: 13%
Concentration Lithium diisopropylamide: 26.0% (2.0M)
Fieser 02,49; 03,184; 04,298; 05,400; 06,334; 07,204; 08,292; 09,280; 10,241; 11,296; 12,277; 14,193; 15,188; 16,15; 17,165
Density 0.8100g/mL
Merck Index 15, 3215
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in thf, n-heptane, ethylbenzene, miscible with liquid aliphatic and aromatic, hydrocarbons, ethers and tertiary amines
Formula Weight 107.12
Percent Purity 24.6 to 30.0% (Lithium diisopropylamide)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.81
Quantity 100 mL
Flash Point 2°C
Physical Form Solution
Chemical Name or Material Lithium diisopropylamide, 2M Solution in THF/n-Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium diisopropylamide, 2M solution in THF/n-heptane/ethylbenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.