missing translation for 'onlineSavingsMsg'
Learn More

Lithium solution for ICP 1g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12966194 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12966194 250 mL Bottle 250mL
12956194 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12966194 Supplier Fisher Chemical Supplier No. S 130 250
  • ONLINE EXCLUSIVE PRICE
    98.55 €
    83.72 € / 250mL
    Save 14.83 €
    15% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Li
Concentration 1±0.002 g/L
Type Mass Spectrometry Standard
Element Li
Solvent or Matrix Nitric Acid
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.