missing translation for 'onlineSavingsMsg'
Learn More

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl

Product Code. 10286680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10286680 100 mL 100mL
10388330 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286680 Supplier Thermo Scientific Acros Supplier No. 196081000
  • ONLINE EXCLUSIVE PRICE
    38.65 €
    37.50 € / 100mL
    Save 1.15 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium standard solution
Name Note 1 mg/ml Li in 2% HCl
Packaging Plastic bottle
CAS 7439-93-2, 7647-01-0, 7732-18-5
Density 0.53
Physical Form Solution
Molecular Formula Li
Concentration 995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C)
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
SMILES [Li]
IUPAC Name lithium
Molecular Weight (g/mol) 6.94
PubChem CID 3028194
Quantity 100 mL
ChEBI CHEBI:30145
Formula Weight 6.94
Grade AAS
Show More Show Less
compliance-icons
Product Identifier
  • Lithium atomic absorption standard solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.