missing translation for 'onlineSavingsMsg'
Learn More

Lithium triethylborohydride, 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 11369018
Change view
Click to view available options
Quantity:
25 mL
50 mL
100 mL
Unit Size:
100mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369018 25 mL 25mL
15444875 50 mL 50mL
11359018 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11369018 Supplier Thermo Scientific Alfa Aesar Supplier No. 042491.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-16-3
Molecular Formula C6H16BLi
Molecular Weight (g/mol) 105.94
MDL Number MFCD00011703
InChI Key WCJAYABJWDIZAJ-UHFFFAOYSA-N
Synonym Lithium triethylhydridoborate
IUPAC Name lithium(1+) triethylboranuide
SMILES [Li+].CC[BH-](CC)CC

Specifications

Color Colorless
Density 0.892g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula LiB(CH2CH3)3H
Quantity 25 mL
UN Number UN3399
Beilstein 4151240
Sensitivity Moisture Sensitive
Merck Index 14,5544
Formula Weight 105.94
Concentration or Composition (by Analyte or Components) 1M in THF
Physical Form Liquid
Chemical Name or Material Lithium triethylborohydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.