missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Liver Tissue Slides (Diabetes)- Frozen

Product Code. 18233384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18233384 5 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18233384 Supplier R&D Systems™ Supplier No. NBP259070
  • ONLINE EXCLUSIVE PRICE
    951.49 €
    845.25 € / Pack of 5
    Save 106.24 €
    11% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Frozen tissue slides are ideal for rapidly identifying cellular localization of RNA or protein.

  • Species Human
  • Type: Tissue
  • Tissues: Liver
  • Tissue Condition: Diabetes
TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Type Liver Tissue Slides
Quantity 5
Regulatory Status RUO
Specimen Type Tissue
Tissue Condition Diabetes
Product Line Novus

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.