missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMP1 Antibody (S20-D), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-79009-100ul
This item is not returnable.
View return policy
Description
LMP1 Monoclonal specifically detects LMP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LMP1 | |
| Monoclonal | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| BNLF1, Latent membrane protein 1, LMP1, LMP-1, Protein p63 | |
| Peptide derived from the C-terminal region of Epstein-Barr virus, Latent Membrane Protein-1 | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S20-D | |
| Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100-1:200 | |
| P03230 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3783750 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction