missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMP744 hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Description
LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity.
Specifications
Specifications
| Chemical Name or Material | LMP744 hydrochloride |
| CAS | 308246-57-3 |
| Color | Dark Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C24H25ClN2O7 |
| Quantity | 5 mg |
| Synonym | MJ-III65 hydrochlorideNSC706744 hydrochloride |
| Solubility Information | DMSO : 105 mg/mL (214.76 mM; Need ultrasonic) |
| SMILES | OCCNCCCN1C2=C(C(C3=CC(OCO4)=C4C=C23)=O)C5=CC(OC)=C(OC)C=C5C1=O.Cl |
| Molecular Weight (g/mol) | 488.92 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction