missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMX1b Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41194
This item is not returnable.
View return policy
Description
LMX1b Polyclonal specifically detects LMX1b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LMX1b | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LMX1B | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human LMX1B. The immunogen is located within amino acids 280 - 330 of LMX1B. Amino Acid Squence: CRLGQGEPGPGQGLGQEV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms are known to exist. This antibody is predicted to not cross-react with LMX1A. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 - 2 ug/ml, ELISA | |
| LIM homeobox transcription factor 1, beta, LIM homeobox transcription factor 1-beta, LIM/homeobox protein 1.2, LIM/homeobox protein LMX1B, LMX1.2, LMX-1.2, MGC138325, MGC142051, NPS1 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mg | |
| Neuroscience | |
| 4010 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction