missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LNX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720004
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Goat, Bovine, Horse, Pig, Cat and Rabbit.
LNX2 (Ligand of Numb Protein X 2) protein is encoded by the LNX2 gene located on chromosome 13 in humans and is known as PDZRN1 (PDZ domain-containing RING finger 1). It is through its PDZ domains that LNX2 forms oligomers and interacts with some of its target proteins. LNX2 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus modulates the Notch signaling pathway.
Specifications
| LNX2 | |
| Polyclonal | |
| Unconjugated | |
| LNX2 | |
| 9630046H24; AW209022; ligand of Numb protein X 2; ligand of numb-protein X 2; Lnx2; Numb-binding protein 2; PDZ domain containing ring finger 1; PDZ domain-containing RING finger protein 1; PDZRN1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140887, 222484, 360761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| Q8N448, Q91XL2 | |
| LNX2 | |
| Multiple peptides against Human LNX2 (aa 442-458, 129-145, 671-683). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction