missing translation for 'onlineSavingsMsg'
Learn More

Lodoxamide tromethamine, MedChemExpress

Product Code. 16439567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439567 Supplier Medchem Express Supplier No. HY16289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lodoxamide tromethamine (U-42585E) is a medication for the treatment of prophylaxis of mast cell-mediated allergic disease.

Lodoxamide tromethamine (U-42585E) is a medication for the treatment of prophylaxis of mast cell-mediated allergic disease.

Specifications

Chemical Name or Material Lodoxamide tromethamine
Physical Form Solid
CAS 63610-09-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H28ClN5O12
For Use With (Application) Neuroscience-Neuromodulation
Synonym U-42585E
Solubility Information DMSO : 20 mg/mL (36.11 mM; Need ultrasonic and warming)
SMILES N#CC1=CC(NC(C(O)=O)=O)=C(Cl)C(NC(C(O)=O)=O)=C1.OCC(CO)(N)CO.OCC(CO)(N)CO
Molecular Weight (g/mol) 553.9
Formula Weight 553.9
Percent Purity 95.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.