missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine hydrochloride, MedChemExpress

Product Code. 16424537
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424537 Supplier Medchem Express Supplier No. HYB1052
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lofexidine (hydrochloride) is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal.

Lofexidine (hydrochloride) is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal.

Specifications

Chemical Name or Material Lofexidine hydrochloride
CAS 21498-08-8
Color White
Purity Grade Notes Research
Molecular Formula C11H13Cl3N2O
Quantity 50 mg
Synonym Baq-168 MDL-14042
Solubility Information H2O : ≥ 100 mg/mL (338.31 mM) ∣DMSO : 100 mg/mL (338.31 mM; Need ultrasonic)
SMILES CC(C1=NCCN1)OC2=C(Cl)C=CC=C2Cl.[H]Cl
Molecular Weight (g/mol) 295.59
Formula Weight 295.59
Percent Purity 98.36%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.