missing translation for 'onlineSavingsMsg'
Learn More

Lomefloxacin hydrochloride, MedChemExpress

Product Code. 16468316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468316 100 mg 100mg
16412146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468316 Supplier Medchem Express Supplier No. HYB0455
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lomefloxacin (SC47111A) hydrochloride is a broad-spectrum quinolone antibiotic, with antimicrobial activity. Lomefloxacin hydrochloride is used for the research of respiratory tract infections, genitourinary infections, gastrointestinal infections, ENT infections, etc..

Lomefloxacin (SC47111A) hydrochloride is a broad-spectrum quinolone antibiotic, with antimicrobial activity. Lomefloxacin hydrochloride is used for the research of respiratory tract infections, genitourinary infections, gastrointestinal infections, ENT infections, etc..

Specifications

Chemical Name or Material Lomefloxacin hydrochloride
Physical Form Solid
CAS 98079-52-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H20ClF2N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym SC47111A hydrochloride
Solubility Information H2O : 10 mg/mL (25.79 mM; Need ultrasonic and warming)
SMILES CCN(C=C(C(O)=O)C1=O)C(C1=CC(F)=C2N(CCN3)CC3C)=C2F.Cl
Molecular Weight (g/mol) 387.81
Formula Weight 387.81
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.