missing translation for 'onlineSavingsMsg'
Learn More

Lomerizine dihydrochloride, MedChemExpress

Product Code. 16444477
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444477 Supplier Medchem Express Supplier No. HYB0768A
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lomerizine dihydrochloride is an antagonist of L- and T-type voltagegated calcium channels.

Lomerizine dihydrochloride is an antagonist of L- and T-type voltagegated calcium channels.

Specifications

Chemical Name or Material Lomerizine dihydrochloride
CAS 101477-54-7
Color White
Purity Grade Notes Research
Molecular Formula C27H32Cl2F2N2O3
Quantity 50 mg
Synonym KB-2796
Solubility Information DMSO : 125 mg/mL (230.86 mM; Need ultrasonic)
SMILES COC1=CC=C(CN2CCN(C(C3=CC=C(F)C=C3)C4=CC=C(F)C=C4)CC2)C(OC)=C1OC.[H]Cl.[H]Cl
Molecular Weight (g/mol) 541.46
Formula Weight 541.46
Percent Purity 99.82%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.