Learn More
Invitrogen™ Loncastuximab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559467
 View more versions of this product
Product Code. 30283301
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Loncastuximab is a biosimilar that targets CD19 .
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Loncastuximab Chimeric
Recombinant Monoclonal
Unconjugated
RB4v1.2
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized