Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
Beskrivelse
- Does not contain any lymphokines required for activation of human lymphocytes
- Intended for in vitro procedures
- Device Master Records (DMRs) for these products are held in an FDA Master File
- Chemically defined media optimized for specific cell types
- For further manufacturing or laboratory use
- Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
- Limited by Federal (USA) law to investigational use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Tekniske data
Tekniske data
| Product Type | Stem Cell Serum Free Media (SFM) |
| With Additives | Phenol Red |
| Quantity | 1L |
| Content And Storage | 2° to 8°C |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?