missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ X-VIVO™ 20 Hematopoietic Serum-Free Culture Media

Product Code. 11615130
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11615130 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11615130 Supplier Lonza™ Supplier No. 04448Q
  • ONLINE EXCLUSIVE PRICE
    330.00 €
    274.47 € / 1 L
    Save 55.53 €
    16% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For adoptive immunotherapy and cellular therapy research

  • Does not contain any lymphokines required for activation of human lymphocytes
  • Intended for in vitro procedures
  • Chemically defined media optimized for specific cell types
  • For further laboratory use
  • Developed and optimized to support the generation of LAK cells from monocyte-depleted PBL at high density
  • Limited by Federal (USA) law to investigational use
  • These products are for research use only
  • Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Serum Free Media (SFM)
With Additives Gentamicin, Phenol Red
Quantity 1 L
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.