missing translation for 'onlineSavingsMsg'
Learn More

Losartan (D4 Carboxylic Acid), MedChemExpress

Product Code. 16448047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16448047

Brand: Medchem Express HY12765S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Losartan D4 Carboxylic Acid (E-3174 D4) is the deuterium labeled Losartan(EXP-3174), which is an angiotensin II receptor antagonist.

Losartan D4 Carboxylic Acid (E-3174 D4) is the deuterium labeled Losartan(EXP-3174), which is an angiotensin II receptor antagonist.

Specifications

Chemical Name or Material Losartan (D4 Carboxylic Acid)
CAS 1246820-62-1
Purity Grade Notes Research
Physical Form Powder
Molecular Formula C22H17D4ClN6O2
Color Off-White
Synonym E-3174 D4 EXP-3174 D4
SMILES ClC1=C(C(O)=O)N(CC2=C([2H])C([2H])=C(C3=CC=CC=C3C4=NN=NN4)C([2H])=C2[2H])C(CCCC)=N1
Industry Type Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 440.92
Quantity 5 mg
Formula Weight 440.92
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.