missing translation for 'onlineSavingsMsg'
Learn More

LP-533401, MedChemExpress

Product Code. 16493496
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493496 25 mg 25mg
16486868 10 mg 10mg
16469407 5 mg 5mg
16461559 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493496 Supplier Medchem Express Supplier No. HY15849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LP-533401 is a Tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.

LP-533401 is a Tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.

Specifications

Chemical Name or Material LP-533401
CAS 945976-43-2
Color Brown
Purity Grade Notes Research
Molecular Formula C27H22F4N4O3
Quantity 25 mg
SMILES N[C@@H](CC1=CC=C(C2=NC(N)=NC(OC(C3=CC=C(C4=CC=CC(F)=C4)C=C3)C(F)(F)F)=C2)C=C1)C(O)=O
Molecular Weight (g/mol) 526.48
Formula Weight 526.48
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.