missing translation for 'onlineSavingsMsg'
Learn More

LPCAT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16662715 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16662715 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16662715 Supplier Thermo Scientific Supplier No. PA5106278

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

LPCAT1 Polyclonal Antibody

LPCAT1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Especificaciones

Antigen LPCAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LPCAT1
Gene Accession No. Q1HAQ0, Q3TFD2, Q8NF37
Gene Alias 1-acylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; 2900035H07Rik; acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; acyltransferase-like 2; AGPAT10; AGPAT9; Aytl2; BB137372; BC005662; C87117; LPC acyltransferase 1; LPCAT; LPCAT1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; lysophosphatidylcholine acyltransferase 1; mLPCAT1; PFAAP3; Phosphonoformate immuno-associated protein 3; rd11; regulated by phosphonoformate; RGD1311599
Gene Symbols LPCAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human LPCAT1(Accession Q8NF37), corresponding to amino acid residues F174-A224.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210992, 361467, 79888
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.