missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPHN2 Monoclonal Antibody (1030202)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560896

Product Code. 30533901

  • 419.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The Latrophilin family of G protein-coupled receptors consists of three members, latrophilin-1, 2 and 3, each of which displays distinct tissue distribution and function. Latrophilin-1, the most characterized member of this family, acts as a receptor for a-latrotoxin, a component of venom from the black widow spider. Binding of a-latrotoxin to latrophilin-1 triggers synaptic vesicle exocytosis via both Ca2+-dependent and -independent mechanisms, resulting in vesicle depletion. Latrophilin-1 is abundant in brain and present in endocrine cells. Latrophilin-3 is also brain-specific, whereas latrophilin-2 expression is ubiquitous.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPHN2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O95490
ADGRL2
Human Latrophilin 2/LPHN2 synthetic peptide.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot
1030202
Unconjugated
ADGRL2
Adgrl2; Adhesion G protein-coupled receptor L2; AI450192; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; Cirl2; Cirl-2; Cl2; Cl2aa; Cl2ac; Gm619; KIAA0786; latrophilin 1; latrophilin 2; latrophilin homolog 1; Latrophilin-2; LEC1; Lectomedin-1; LPHH1; Lphn2; mKIAA0786
Mouse
Protein A/G
RUO
23266
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.