missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPL Recombinant Rabbit Monoclonal Antibody (10E10P9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535444
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: IPFTLPEVST NKTYSFLIYT EVDIGELLML KLKWKSDSYF SWSDWWSSPG FAIQKIRVKA GETQKKVIFC SREKVSHLQK GKAPAVFVKC HDKSLNKKSG.
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifica
| LPL | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06858, P11152 | |
| LPL | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 10E10P9 | |
| Unconjugated | |
| LPL | |
| adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16956, 4023 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto