missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRH-1/NR5A2 Antibody (SR2100), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21744-50ul
This item is not returnable.
View return policy
Description
LRH-1/NR5A2 Monoclonal antibody specifically detects LRH-1/NR5A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| LRH-1/NR5A2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2100 | |
| Western Blot 1:500-1:2000 | |
| Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
| A synthesized peptide derived from human LRH-1/NR5A2 (Uniprot #: O00482) | |
| 50 μg | |
| Cancer, Chromatin Research, Neuroscience, Stem Cell Markers | |
| 2494 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction