missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRIG1 Monoclonal Antibody (440710)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561337
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
LRIG1 protein Q96JA1 is coded by a gene on a chromosome band 3p14.3, this region is known to be deleted in various human cancers. LRIG1 is considered to be a tumor suppressor gene in humans. LRIG1 is an integral cell-surface membrane protein that is expressed by specific cells in various human tissues and that its 143-kDa form might be cleaved into 111-kDa and 32-kDa fragments. The LRIG1 protein may inhibit the growth of tumors of glial cells and the down-regulation of the LRIG1 gene may be involved in the development and progression of the tumor.
Specifications
| LRIG1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P70193 | |
| Lrig1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LRIG1, Ala37-Thr794. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Inhibition Assays | |
| 440710 | |
| Unconjugated | |
| Lrig1 | |
| D6Bwg0781e; DKFZp586O1624; Img; integral membrane glycoprotein; leucine rich repeats and immunoglobulin like domains 1; leucine-rich repeat protein LRIG1; leucine-rich repeats and immunoglobulin like domains 1; leucine-rich repeats and immunoglobulin-like domains 1; leucine-rich repeats and immunoglobulin-like domains protein 1; LIG1; LIG-1; LRIG1; ortholog of mouse integral membrane glycoprotein LIG-1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 16206 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction