missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-6, p Ser1490 Antibody (SR1321), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22610-50ul
This item is not returnable.
View return policy
Description
LRP-6 Monoclonal antibody specifically detects LRP-6 in Human samples. It is validated for Western Blot
Specifications
| LRP-6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1321 | |
| Western Blot 1:500-1:2000 | |
| ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
| A synthesized peptide derived from human phospho-LRP-6 [Ser1490] (Uniprot #: O75581) | |
| 50 μg | |
| Phospho Specific, Stem Cell Signaling Pathway | |
| 4040 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction