missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32/GARP Antibody (GARP5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37710H
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Human samples. It is validated for Western Blot
Specifications
| LRRC32/GARP | |
| Monoclonal | |
| HRP | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| Purified human sGARP protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| GARP5 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2615 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction