missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRC4 Monoclonal Antibody (701419)
GREENER_CHOICE

Product Code. 30535760
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535760 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535760 Supplier Invitrogen™ Supplier No. MA561507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The LRRC4 gene, also known as netrin-G ligand 2 (NGL-2), is a brain-specific member of the leucine-rich repeat superfamily, playing a critical role in the central nervous system. Its expression is particularly high in areas such as the hippocampal CA1 and CA2 regions, as well as in granule cells of the dentate gyrus. LRRC4 is a known tumor suppressor gene associated with glioma suppression, as it regulates glioma cell growth, angiogenesis, and invasion through extensive signaling networks. It can participate in multiphase loops involving miRNA, transcription factors, and gene methylation modifications. This gene targets various genes involved in cell motility, impacting glioblastoma cell movement and polarization by binding to enhancer elements and activating transcription, notably the RAP1GAP gene. Its function and expression are crucial in maintaining normal brain function and hold potential implications in the management of gliomagenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 701419
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Lrrc4
Gene Accession No. Q9HBW1
Gene Alias BAG; brain tumor associated protein LRRC4; brain tumor-associated protein BAG; Brain tumor-associated protein MBAG1; leucine rich repeat containing 4; leucine rich repeat containing 4 protein; leucine-rich repeat-containing 4; leucine-rich repeat-containing protein 4; LIBG-like protein; LRRC4; LRRC4-like protein; MBAG1; NAG14; Nasopharyngeal carcinoma-associated gene 14 protein; netrin-G2 ligand; NGL-2; NLG2; NLG-2; protein NGL-2; UNQ554/PRO1111
Gene Symbols Lrrc4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRRC4, Ala39-Lys527.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.