missing translation for 'onlineSavingsMsg'
Learn More

LRRK2 Antibody (SR1449), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00 € - 449.00 €

Specifications

Antigen LRRK2
Clone SR1449
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18679324
View Documents
Novus Biologicals
NBP3-21626-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18693474
View Documents
Novus Biologicals
NBP3-21626-50ul
50 μg
338.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

LRRK2
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2
A synthesized peptide derived from human LRRK2 (Uniprot #: Q5S007)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1449
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
120892
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.