missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSAMP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149485
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system. During development of the limbic system, this encoded protein is found on the surface of axonal membranes and growth cones, where it acts as a selective homophilic adhesion molecule, and guides the development of specific patterns of neuronal connections.
Specifications
| LSAMP | |
| Polyclonal | |
| Unconjugated | |
| Lsamp | |
| 5430428I19; AW046674; B130007O04Rik; D930023J12Rik; IgLON family member 3; IGLON3; Lam; LAMP; limbic system associated membrane protein; limbic system-associated membrane protein; Lsamp; neuronal growth regulator 1-like; Unknown (protein for MGC:128495) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4045 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q13449 | |
| Lsamp | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP, Val29-Asn315. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction