missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTA4H Monoclonal Antibody (24GB5540)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557443
This item is not returnable.
View return policy
Description
LTA4H Monoclonal Antibody for Western Blot, Flow
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications
| LTA4H | |
| Monoclonal | |
| Unconjugated | |
| LTA4H | |
| Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 16993, 299732, 4048 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5540 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09960, P24527, P30349 | |
| LTA4H | |
| Recombinant protein of human LTA4H (aa 1-611). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction