missing translation for 'onlineSavingsMsg'
Learn More

LTF Mouse, anti-Human, Clone: 3D1G3, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen LTF
Clone 3D1G3
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18039815
View Documents
Proteintech
68509-1-IG-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18028115
View Documents
Proteintech
68509-1-IG-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The LTF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LTF. This antibody recognizes human antigen. The LTF antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

LTF
1000 μg/mL
Liquid
Human
GIG12, HLF2, Lactoferrin, lactotransferrin, LF, LTF, Talalactoferrin
Primary
3D1G3
Western Blot, ELISA
Mouse
4057
IgG2a
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.