missing translation for 'onlineSavingsMsg'
Learn More

Lubricating Oil Standard 14: Ca @ 0.010; P @ 0.125; S @ 0.600; Zn @ 0.060 wt% in Lubricating Oil, VHG™

Product Code. 17983076
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983076 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17983076 Supplier VHG Labs Supplier No. VHGLOIL14100
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lubricating Oil Standard 14: Ca @ 0.010; P @ 0.125; S @ 0.600; Zn @ 0.060 wt% in Lubricating Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Zinc/Phosphorus/Sulfur
CAS 7440-70-2, 7440-66-6, 7723-14-0, 7704-34-9
Methods XRF
Analyte or Component Names Calcium/Zinc/Phosphorus/Sulfur
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 20 cSt Mineral Oil
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Calcium 0.01%/Zinc 0.06%/Phosphorus 0.125%/Sulfur 0.5999%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.