missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LUC7L2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00 € - 470.00 €
Specifications
| Antigen | LUC7L2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18368734
|
Novus Biologicals
NBP3-04955-20UL |
20 μL |
193.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18321846
|
Novus Biologicals
NBP3-04955-100UL |
100 μL |
470.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LUC7L2 Polyclonal specifically detects LUC7L2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| LUC7L2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 51631 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CGI-59, CGI-74, FLJ10657, H_NH0792N18.3, hLuc7B2, LUC7B2, LUC7-like 2 (S. cerevisiae), putative RNA-binding protein Luc7-like 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 313-392 of human LUC7L2 (NP_057103.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title