missing translation for 'onlineSavingsMsg'
Learn More

Lumefantrine, MedChemExpress

Product Code. 16448466
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448466 100 mg 100mg
16442216 500 mg 500mg
16428628 10 mg 10mg
16424497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448466 Supplier Medchem Express Supplier No. HYB0803
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lumefantrine is an antimalarial drug, used in combination with Artemether.

Lumefantrine is an antimalarial drug, used in combination with Artemether.

Specifications

Chemical Name or Material Lumefantrine
Physical Form Solid
CAS 82186-77-4
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C30H32Cl3NO
For Use With (Application) COVID-19-immunoregulation
Synonym Benflumetol
Solubility Information DMF : 25 mg/mL (47.26 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 2 mg/mL (3.78 mM; Need ultrasonic)
SMILES OC(CN(CCCC)CCCC)C1=CC(Cl)=CC(/C2=C\C3=CC=C(Cl)C=C3)=C1C4=C2C=C(Cl)C=C4
Molecular Weight (g/mol) 528.94
Formula Weight 528.94
Percent Purity 98.41%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.