missing translation for 'onlineSavingsMsg'
Learn More

Lumefantrine, TRC

Product Code. 17930555
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930555 25 mg 25mg
17912485 100 mg 100mg
17922485 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930555 Supplier Toronto Research Chemicals Supplier No. TRCL47400025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82186-77-4
Molecular Formula C30 H32 Cl3 N O
Molecular Weight (g/mol) 528.94
Synonym 9H-Fluorene-4-methanol, 2,7-dichloro-9-[(4-chlorophenyl)methylene]-α-[(dibutylamino)methyl]-, (9Z)- (9CI), 9H-Fluorene-4-methanol, 2,7-dichloro-9-[(4-chlorophenyl)methylene]-α-[(dibutylamino)methyl]-, (Z)-, Benflumelol, Benflumetol, Lumefantrine, dl-Benflumelol
IUPAC Name 2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]fluoren-4-yl]ethanol
SMILES CCCCN(CCCC)CC(O)c1cc(Cl)cc2\C(=C/c3ccc(Cl)cc3)\c4cc(Cl)ccc4c12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H32Cl3NO/c1-3-5-13-34(14-6-4-2)19-29(35)28-18-23(33)17-27-25(15-20-7-9-21(31)10-8-20)26-16-22(32)11-12-24(26)30(27)28/h7-12,15-18,29,35H,3-6,13-14,19H2,1-2H3/b25-15-
Formula Weight 527.1549
Chemical Name or Material Lumefantrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.