missing translation for 'onlineSavingsMsg'
Learn More

Lumibudesonide, TRC

Product Code. 30477902
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477902 10 mg 10mg
30478883 5 mg 5mg
30468948 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477902 Supplier Toronto Research Chemicals Supplier No. TRCL48966010MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lumibudesonide
Analyte or Component Names Lumibudesonide
Shipping Condition Room Temperature
Molecular Formula C25 H34 O6
Synonym (5aR,5bS,5cS,6S,7aS,7bS,10aR,11aS,11bS)-6-hydroxy-7b-(2-hydroxyacetyl)-5b,7a-dimethyl-9-propyl-1,5a,5b,5c,6,7,7a,7b,10a,11,11a,11b-dodecahydrocyclopenta[2'',3'']cyclopropa[1'',2'':3',4']benzo[1',2':4,5]indeno[1,2-d][1,3]dioxol-5(2H)-one
InChI Formula InChI=1S/C25H34O6/c1-4-5-19-30-18-10-14-13-6-8-24-9-7-15(27)21(24)23(24,3)20(13)16(28)11-22(14,2)25(18,31-19)17(29)12-26/h7,9,13-14,16,18-21,26,28H,4-6,8,10-12H2,1-3H3/t13-,14-,16-,18+,19?,20+,21-,22-,23+,24+,25+/m0/s1
Solution Type Neat
SMILES CCCC1O[C@@H]2C[C@H]3[C@@H]4CC[C@@]56C=CC(=O)[C@H]5[C@@]6(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC Name (1S,2S,4R,8S,9S,11S,12S,13R,14S,18R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxahexacyclo[10.8.0.02,9.04,8.013,18.014,18]icos-16-en-15-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 430.534
Quantity 10 mg
Formula Weight 430.236
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.