missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lumiliximab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
477.00 € - 1204.00 €
Specifications
| Antigen | Lumiliximab Chimeric |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284424
|
Invitrogen™
MA559423 |
100 μg |
477.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283542
|
Invitrogen™
MA559424 |
1 mg |
1204.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lumiliximab is a chimeric monoclonal antibody which is used as an immunosuppressive pharmaceutical. It is a primatized anti-CD23 macaque/human chimeric antibody that inhibits the production of the IgE antibody, for the potential treatment of allergic conditions. Lumiliximab was developed by IDEC Pharmaceuticals, which was acquired by Biogen. Clinical trials for CLL were terminated in 2010, and for allergic asthma in 2007. Results published from the CLL clinical trial failed to meet primary endpoints. Lumiliximab is a chimeric macaque-human monoclonal antibody to CD23, a protein expressed on virtually all chronic lymphocytic leukemia (CLL) cells. CD23, also known as Fc epsilon RII, or FcepsilonRII, is the 'low affinity' receptor for IgE, an antibody isotype involved in allergy and resistance to parasites and is important in regulation of IgE levels. Unlike many of the antibody receptors, CD23 is a C-type lectin. It is found on mature B cells, activated macrophages, eosinophils, follicular dendritic cells and platelets.Specifications
| Lumiliximab Chimeric | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| IDEC-152; P5E8 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title