missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lung Matched Tumor and Normal Tissue Lysate

Product Code. 18613096 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vial
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613096 2 Vial Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613096 Supplier Novus Biologicals™ Supplier No. NBP2470952Vial
  • ONLINE EXCLUSIVE PRICE
    685.00 €
    647.85 € / Pack of 2
    Save 37.15 €
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tissue Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method These lysates have not been subjected to denaturing or reducing conditions. This allows the tissue or cell lysate to be used in a variety of applications; to study protein-protein interaction, ligand binding, ELISA, immunoprecipitation, 1 D and 2 D gel electrophoresis, and Western blotting for the detection of specific protein targets. For use in 1 D and 2 D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required. Buffer requirements for performing protein-protein interaction and ligand binding studies can vary significantly from RIPA buffer and may require modifications. In most cases, tissue lysates in RIPA buffer can be used, directly in standard ELISA and immunoprecipitation assays. These lysates are proteomic discovery tools. Researchers should validate and optimize for individual use.
Quantity 2 Vial
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target Lung
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.