missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteolin-7-O-glucuronide, Dr. Ehrenstorfer
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Luteolin- 7-O-Glucuronide |
| CAS | 29741-10-4 |
| Analyte or Component Names | Luteolin- 7-O-Glucuronide |
| Shipping Condition | +20°C |
| Molecular Formula | C21H18O12 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Cyanidenon-7-O-beta-D-glucuronic Acid, Luteolin 7-O-Glucuronide, Luteolin 7-O-beta-D-Glucuronide, Luteolin 7-O-beta-D-Glucuronopyranoside, Luteolin 7-O-beta-Glucuronide, Luteolin 7-O-beta-Glucuronopyranoside, Luteolin 7-Glucuronide;Luteolin-7-beta-D-Glucuronide |
| SMILES | O[C@@H]1[C@@H](O)[C@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)c(O)c4)O[C@@H]([C@H]1O)C(=O)O |
| Molecular Weight (g/mol) | 462.36 |
| Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction