missing translation for 'onlineSavingsMsg'
Learn More

LW6, MedChemExpress

Product Code. 16495798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495798 10 mg 10mg
16473376 25 mg 25mg
16421957 50 mg 50mg
16408477 5 mg 5mg
16406276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495798 Supplier Medchem Express Supplier No. HY13671
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.

LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.

Specifications

Chemical Name or Material LW6
CAS 934593-90-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29NO5
Quantity 10 mg
Synonym HIF-1α inhibitor LW8
Solubility Information DMSO : 25 mg/mL (57.40 mM; Need ultrasonic) ∣DMF : 17.24 mg/mL (39.59 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(O)C(NC(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)=C1
Molecular Weight (g/mol) 435.51
Formula Weight 435.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.