missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Red 685, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

185.00 € - 348.00 €

Specifications

Antigen Ly-108
Clone eBio13G3-19D (13G3-19D)
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055474
View Documents
Invitrogen™
M050T03R02-A
100 μg
348.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055015
View Documents
Invitrogen™
M050T02R02-A
25 μg
185.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ly-108 Monoclonal antibody specifically detects Ly-108 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

Ly-108
0.1 mg/mL
Monoclonal
Mouse
Mouse
Q9ET39
30925
Primary
4°C, store in dark, DO NOT FREEZE!
eBio13G3-19D (13G3-19D)
Flow Cytometry
NovaFluor Red 685
RUO
PBS with 0.09% sodium azide; pH 7.2
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080
IgG2a κ
Affinity chromatography
SLAMF6
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.